Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGTTTTGGAACTTCTGCCCTCC[A/G]CTGATGTGTTTATACAAAGAAGCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Phactr2 PubMed Links |
Phactr2 - phosphatase and actin regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033257.4 | Intron | NP_001028429.1 | ||||
NM_001195065.1 | Intron | NP_001181994.1 | ||||
NM_001195066.1 | Intron | NP_001181995.1 | ||||
NM_001195096.1 | Intron | NP_001182025.1 |