Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCAGCAGGTGACAAGTTAGCCC[A/G]TTTGTATGTTAAGGTCACAGAAAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mta3 PubMed Links |
Mta3 - metastasis associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171052.1 | Intron | NP_001164523.1 | ||||
NM_001171053.1 | Intron | NP_001164524.1 | ||||
NM_001171054.1 | Intron | NP_001164525.1 | ||||
NM_054082.2 | Intron | NP_473423.1 |