Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCATCATGTCCCACCAGGCAAT[A/G]CCGGCTGACAGTTTGAGAAAAATCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Anks1b PubMed Links |
Anks1b - ankyrin repeat and sterile alpha motif domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128086.2 | Intron | NP_001121558.2 | ||||
NM_001177396.1 | Intron | NP_001170867.1 | ||||
NM_001177397.1 | Intron | NP_001170868.1 | ||||
NM_001177398.1 | Intron | NP_001170869.1 | ||||
NM_181398.3 | Intron | NP_852063.1 |