Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTAAACACTAAGATACTCCACTT[A/T]AAAAAAAATAATCTCCAGGATGGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ust PubMed Links |
Ust - uronyl-2-sulfotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177387.3 | Intron | NP_796361.2 |