Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTACTCTGAAAATAATAATGTC[A/G]AAAGATTTAATAGCCCATATTGTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Csmd3 PubMed Links |
Csmd3 - CUB and Sushi multiple domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081391.2 | Intron | NP_001074860.2 |