Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATATAGGATAGTTTTAGAAGAA[C/G]TTCTGGTTATCAGTCACTATTCTGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cpne3 PubMed Links |
Cpne3 - copine III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027769.2 | Intron | NP_082045.1 |