Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTCCTGGAAAAAGACACTGTAT[A/G]AACAGGCCTCCTTCCCTTTCATTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rnf38 PubMed Links |
Rnf38 - ring finger protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038993.3 | Intron | NP_001034082.1 | ||||
NM_175201.5 | Intron | NP_780410.2 |