Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Actl7a PubMed Links |
Actl7a - actin-like 7a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009611.3 | 1160 | Silent Mutation | NP_033741.1 |
Actl7b - actin-like 7b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Ikbkap - inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |