Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACACCAAAACCACAGTGTGATA[C/A]CAACCAACCAGGCCAGAGCAGGCCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Wdr91 PubMed Links |
Wdr91 - WD repeat domain 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013366.1 | Intron | NP_001013384.1 |