Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATTCTTTTTCGGAAGGAACCAG[A/C]AATTACAATTCTCTCAGCAATTCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Wdr70 PubMed Links |
Wdr70 - WD repeat domain 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081402.1 | Intron | NP_001074871.1 |