Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGATGATTCTGAGTGATAGACA[C/A]TAACATAAAGCAAGTGTCTTCAAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trps1 PubMed Links |
Trps1 - trichorhinophalangeal syndrome I (human) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032000.2 | Intron | NP_114389.2 |