Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAGGGTCACTGCTGAATTGTCT[C/T]TTTTTCCCCCAGAGTCTGGTCTTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Smoc2 PubMed Links |
Smoc2 - SPARC related modular calcium binding 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022315.2 | Intron | NP_071710.2 |