Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCCCTGGGATGTCCGTTAGCG[C/T]GTTCGACCTTATGCAAAGTGAACGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hrh4 PubMed Links |
Hrh4 - histamine receptor H4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153087.2 | Intron | NP_694727.1 |