Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Garnl3 PubMed Links |
Garnl3 - GTPase activating RANGAP domain-like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178888.4 | 3234 | UTR 3 | NP_849219.2 |
Slc2a8 - solute carrier family 2, (facilitated glucose transporter), member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |