Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Acvr2a PubMed Links |
Acvr2a - activin receptor IIA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Orc4 - origin recognition complex, subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177313.1 | 2218 | Intron | NP_001170784.1 | |||
NM_011958.3 | 2218 | UTR 3 | NP_036088.3 |