Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTAAGTCTTCAAACATTTCGTTG[C/T]TCGTTTTATGCCCTCATGAGCTCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sfrp2 PubMed Links |
Sfrp2 - secreted frizzled-related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009144.2 | 1659 | UTR 3 | NP_033170.1 |