Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAGCACATCAAGCAGGGGCTC[C/T]TGCAGCTTCTCCACCGCCTCTCTGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nr1h4 PubMed Links |
Nr1h4 - nuclear receptor subfamily 1, group H, member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163504.1 | 1302 | Silent Mutation | NP_001156976.1 | |||
NM_001163700.1 | 1302 | Silent Mutation | NP_001157172.1 | |||
NM_009108.2 | 1302 | Silent Mutation | NP_033134.2 |