Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAGTGTAAAATCGTGAGGAGAC[A/G]ATTTCAGTATTCGATATGGTGTGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mboat2 PubMed Links |
Mboat2 - membrane bound O-acyltransferase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083341.1 | 2087 | UTR 3 | NP_001076810.1 | |||
NM_026037.3 | 2087 | UTR 3 | NP_080313.2 |