Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATTAGTATGTGGCCTTTCTACC[A/G]TCAAAGTCCCTGTTGTCATGGTCTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pcsk5 PubMed Links |
Pcsk5 - proprotein convertase subtilisin/kexin type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163144.1 | Intron | NP_001156616.1 | ||||
NM_001190483.1 | Intron | NP_001177412.1 |