Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Abcb8 PubMed Links |
Abcb8 - ATP-binding cassette, sub-family B (MDR/TAP), member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029020.2 | 2688 | UTR 3 | NP_083296.2 |
Asic3 - acid-sensing (proton-gated) ion channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183000.2 | 2688 | Intron | NP_892045.2 |
Cdk5 - cyclin-dependent kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007668.3 | 2688 | Intron | NP_031694.1 |