Search
Search

| Assay Name: | hsa-miR-504-5p |
|---|---|
| miRBase Accession Number: | MI0003189 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAUC |
| Species: | Human, Mouse, Dog, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | QuantiGene Singleplex miRNA |
Gene Family ID: | MIPF0000437, mir-504 |
Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-504-5p | MIMAT0002875 |
| Mouse | mmu-miR-504-5p | MIMAT0004889 |
| Dog | cfa-miR-504 | MIMAT0009907 |
| Guinea pig | cpo-miR-504-5p | MIMAT0047309 |
| Horse | eca-miR-504 | MIMAT0013228 |
| Pan troglodytes | ptr-miR-504 | MIMAT0008177 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-504 | MIMAT0015927 |
| Rabbit | ocu-miR-504-5p | MIMAT0048501 |
| Rhesus monkey | mml-miR-504-5p | MIMAT0006368 |
| Tupaia chinensis | tch-miR-504 | MIMAT0036605 |