Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          Assay Name: bdi-miR399a
          miRBase Accession Number: MI0011572
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UGCCAAAGGAGAAUUACCCUG
          Species: Brachypodium distachyon
          Product Type: QuantiGene Singleplex miRNA
          Assay ID SM-20365
          Size 200 rxns
          Availability Made To Order
          Catalog # QGSM-200
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000015, MIR399

          Mature miRNA Sequence:

          UGCCAAAGGAGAAUUACCCUG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Brachypodium distachyon bdi-miR399a MIMAT0012187

          Stem-loop Details



          Brachypodium distachyon

          Stem-loop ID bdi-MIR399a
          Stem-loop Accession # MI0011572
          Stem-loop Sequence
          CAGUGCAGUUCUCCUCUGGCAUGGAUGCUAAUGUAGCAGGCCCUGCACAAUGGACUAUGCUAUCACUGUCCCUGCCAAAGGAGAAUUACCCUGCCAGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012187
          miRBase ID: bdi-miR399a
          Mature miRNA Sequence: UGCCAAAGGAGAAUUACCCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          QuantiGene Singleplex miRNA Control : SMC-20365


          miRBase Alias:

          Brevundimonas diminuta: bdi-miR399a (v22)

          Back To Top

          Related Products

          • QuantiGene Singleplex Assay Kit
          • QuantiGene Singleplex HT Assay Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline