Search
Search

| Assay Name: | ppe-miR6265 |
|---|---|
| miRBase Accession Number: | MI0021621 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGAACUUUGACCCGAUUCGCAU |
| Species: | Prunus persica |
| Product Type: | QuantiGene Singleplex miRNA |
Mature miRNA Sequence: | UUGAACUUUGACCCGAUUCGCAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Prunus persica | ppe-miR6265 | MIMAT0027284 |