Search
Search

| Assay Name: | asu-miR-36e-3p |
|---|---|
| miRBase Accession Number: | MI0018540 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACCGGGCUCGAAUCCUUGUC |
| Species: | Ascaris suum |
| Product Type: | QuantiGene Singleplex miRNA Control |
Gene Family ID: | MIPF0000096, mir-36 |
Mature miRNA Sequence: | UCACCGGGCUCGAAUCCUUGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ascaris suum | asu-miR-36e-3p | MIMAT0021448 |