Search
Search

| Assay Name: | rno-miR-3584-5p |
|---|---|
| miRBase Accession Number: | MI0015458 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGAGGAGUCCAGGAGGC |
| Species: | Rat |
| Type: | 1 |
| Detection Label: | Alexa Fluor 546 |
| Product Type: | ViewRNA Cell Plus Probe Set |
Gene Family ID: | MIPF0002006, mir-3584 |
Mature miRNA Sequence: | GGGAGGAGUCCAGGAGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3584-5p | MIMAT0017875 |