Search
Search

| Assay Name: | chi-miR-2411-5p |
|---|---|
| miRBase Accession Number: | MI0030715 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGGAGUGACUGUCAGAUGCAGCCAGC |
| Species: | Goat |
| Type: | 1 |
| Detection Label: | Alexa Fluor 546 |
| Product Type: | ViewRNA Cell Plus Probe Set |
Mature miRNA Sequence: | GUGGAGUGACUGUCAGAUGCAGCCAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-2411-5p | MIMAT0036093 |