Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | cel-miR-44 |
---|---|
miRBase Accession Number: | MI0000015 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGACUAGAGACACAUUCAGCU |
Species: | Caenorhabditis elegans, Ascaris suum, Caenorhabditis brenneri, Caenorhabditis briggsae, Caenorhabditis remanei, Haemonchus contortus, Heligmosomoides polygyrus, Pristionchus pacificus |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000132, mir-44 |
Mature miRNA Sequence: | UGACUAGAGACACAUUCAGCU |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Caenorhabditis elegans | cel-miR-44-3p | MIMAT0000015 | |
Caenorhabditis elegans | cel-miR-45-3p | MIMAT0000016 | |
Ascaris suum | asu-miR-44a-3p | MIMAT0021456 | |
Caenorhabditis brenneri | cbn-miR-44 | MIMAT0029711 | |
Caenorhabditis briggsae | cbr-miR-44 | MIMAT0000469 | |
Caenorhabditis briggsae | cbr-miR-45 | MIMAT0000470 | |
View More Rows | |||
Caenorhabditis remanei | crm-miR-44-3p | MIMAT0011588 | |
Caenorhabditis remanei | crm-miR-45-3p | MIMAT0011578 | |
Haemonchus contortus | hco-miR-45 | MIMAT0023339 | |
Heligmosomoides polygyrus | hpo-miR-45-3p | MIMAT0040507 | |
Pristionchus pacificus | ppc-miR-45 | MIMAT0011640 |