Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10353
          Assay Name: hsa-miR-383-5p
          miRBase Accession Number: MI0000791
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 14853438 - 14853510 [-] on Build GRCh38
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Species: Human, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Dasypus novemcinctus, Dog, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata, Xenopus tropicalis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10353
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000137, mir-383

          Mature miRNA Sequence:

          AGAUCAGAAGGUGAUUGUGGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-383-5p MIMAT0000738
          Bovine bta-miR-383 MIMAT0009309
          Callithrix jacchus cja-miR-383a MIMAT0039368
          Chicken gga-miR-383-5p MIMAT0003363
          Chrysemys picta cpi-miR-383-5p MIMAT0037874
          Dasypus novemcinctus dno-miR-383-5p MIMAT0047868
          View More Rows
          Dog cfa-miR-383 MIMAT0006629
          Guinea pig cpo-miR-383-5p MIMAT0047229
          Horse eca-miR-383 MIMAT0013185
          Monodelphis domestica mdo-miR-383-5p MIMAT0004139
          Ophiophagus hannah oha-miR-383-5p MIMAT0036919
          Pan troglodytes ptr-miR-383 MIMAT0008135
          Pongo pygmaeus ppy-miR-383 MIMAT0015873
          Pteropus alecto pal-miR-383-5p MIMAT0040171
          Python bivittatus pbv-miR-383-5p MIMAT0039046
          Rabbit ocu-miR-383-5p MIMAT0048429
          Rhesus monkey mml-miR-383 MIMAT0006312
          Taeniopygia guttata tgu-miR-383-5p MIMAT0014531
          Xenopus tropicalis xtr-miR-383 MIMAT0003639

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-383
          Stem-loop Accession # MI0000791
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location Chr. 8 - 14853438 - 14853510 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000738
          miRBase ID: hsa-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: Chr. 8 - 14853438 - 14853510 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-383
          Stem-loop Accession # MI0009823
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUCAAUCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009309
          miRBase ID: bta-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-383a
          Stem-loop Accession # MI0031883
          Stem-loop Sequence
          GUCACCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAGCAAGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039368
          miRBase ID: cja-miR-383a
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-383
          Stem-loop Accession # MI0003706
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUAAAUAUUGAGCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003363
          miRBase ID: gga-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-383
          Stem-loop Accession # MI0029563
          Stem-loop Sequence
          AAGUCACCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGAGUAGAUAUUAAGCAGCCACAGCACUGCCUGGUCAGAAAGAGCAAGUGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037874
          miRBase ID: cpi-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-383
          Stem-loop Accession # MI0039099
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUACUAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047868
          miRBase ID: dno-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-383
          Stem-loop Accession # MI0008026
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006629
          miRBase ID: cfa-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-383
          Stem-loop Accession # MI0038763
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUAGAUAUUAAUCAGCCACAACACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047229
          miRBase ID: cpo-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-383
          Stem-loop Accession # MI0012930
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013185
          miRBase ID: eca-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-383
          Stem-loop Accession # MI0005328
          Stem-loop Sequence
          CUCAGAUCAGAAGGUGAUUGUGGCUUUCGGUAGACAUGGAACAGCCACAUCACUGGCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004139
          miRBase ID: mdo-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-383
          Stem-loop Accession # MI0031467
          Stem-loop Sequence
          ACCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGUUUGAUAUGAAACAGCCACAGCACUGCCUGGUCAGAAAGAGCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036919
          miRBase ID: oha-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-383
          Stem-loop Accession # MI0008656
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008135
          miRBase ID: ptr-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-383
          Stem-loop Accession # MI0014931
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015873
          miRBase ID: ppy-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-383
          Stem-loop Accession # MI0032532
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040171
          miRBase ID: pal-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-383
          Stem-loop Accession # MI0030307
          Stem-loop Sequence
          CAGUCACCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGUUUGAUAUGAAACAGCCACAGCACUGCCUGGUCAGAAAGAGCAAGUGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039046
          miRBase ID: pbv-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-383
          Stem-loop Accession # MI0039396
          Stem-loop Sequence
          AGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAGCAAUCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048429
          miRBase ID: ocu-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-383
          Stem-loop Accession # MI0007731
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGGGUGGAUAUUAAUCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006312
          miRBase ID: mml-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-383
          Stem-loop Accession # MI0013744
          Stem-loop Sequence
          CCUGCUCCUCAGAUCAGAAGGUGAUUGUGGCUUUGAGUAAAUAUUAAGCAGCCACAGCACUGCCUGGUCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014531
          miRBase ID: tgu-miR-383-5p
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-383
          Stem-loop Accession # MI0004881
          Stem-loop Sequence
          CUCCUCAGAUCAGAAGGUGAUUGUGGCUUUUAGUAGAUAUUAAGCAGCCACAGCACUGCCUGGUCAGAAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003639
          miRBase ID: xtr-miR-383
          Mature miRNA Sequence: AGAUCAGAAGGUGAUUGUGGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303566_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10353
          TaqMan™ MicroRNA Assay : 000573
          mirVana® miRNA inhibitor : MH10353
          mirVana® miRNA mimic : MC10353
          TaqMan™ Advanced miRNA Assay : 478079_mir


          miRBase Alias:

          Human: hsa-miR-383 (v19)
          Gallus gallus: gga-miR-383 (v19)
          Monodelphis domestica: mdo-miR-383 (v19)
          Trabulsiella guamensis ATCC 49490: tgu-miR-383 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline