Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11021
          Assay Name: hsa-miR-99b-5p
          miRBase Accession Number: MI0000746
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 51692612 - 51692681 [+] on Build GRCh38
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Species: Human, Mouse, Rat, Bovine, Dog, Goat, Guinea pig, Horse, Otolemur garnettii, Pan troglodytes, Pig, Pteropus alecto, Rhesus monkey, Tupaia chinensis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11021
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          CACCCGUAGAACCGACCUUGCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-99b-5p MIMAT0000689
          Mouse mmu-miR-99b-5p MIMAT0000132
          Rat rno-miR-99b-5p MIMAT0000821
          Bovine bta-miR-99b MIMAT0004345
          Dog cfa-miR-99b MIMAT0006607
          Goat chi-miR-99b-5p MIMAT0036314
          View More Rows
          Guinea pig cpo-miR-99b-5p MIMAT0046943
          Horse eca-miR-99b MIMAT0013018
          Otolemur garnettii oga-miR-99b MIMAT0049653
          Pan troglodytes ptr-miR-99b MIMAT0008351
          Pig ssc-miR-99b MIMAT0006018
          Pteropus alecto pal-miR-99b-5p MIMAT0039834
          Rhesus monkey mml-miR-99b-5p MIMAT0006179
          Tupaia chinensis tch-miR-99b-5p MIMAT0036582

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-99b
          Stem-loop Accession # MI0000746
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 19 - 51692612 - 51692681 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000689
          miRBase ID: hsa-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 19 - 51692612 - 51692681 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-99b
          Stem-loop Accession # MI0000147
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 17 - 17830188 - 17830257 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000132
          miRBase ID: mmu-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 17 - 17830188 - 17830257 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-99b
          Stem-loop Accession # MI0000884
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000821
          miRBase ID: rno-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-99b
          Stem-loop Accession # MI0005469
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004345
          miRBase ID: bta-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-99b
          Stem-loop Accession # MI0008003
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGAGUCUGUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006607
          miRBase ID: cfa-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-99b
          Stem-loop Accession # MI0030846
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036314
          miRBase ID: chi-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-99b
          Stem-loop Accession # MI0038611
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCACACACAAGCUCGUGUCUGUGGGUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046943
          miRBase ID: cpo-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-99b
          Stem-loop Accession # MI0012767
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013018
          miRBase ID: eca-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-99b
          Stem-loop Accession # MI0040436
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049653
          miRBase ID: oga-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-99b
          Stem-loop Accession # MI0008891
          Stem-loop Sequence
          GCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008351
          miRBase ID: ptr-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-99b
          Stem-loop Accession # MI0007077
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006018
          miRBase ID: ssc-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-99b
          Stem-loop Accession # MI0032353
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039834
          miRBase ID: pal-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-99b
          Stem-loop Accession # MI0007609
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006179
          miRBase ID: mml-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-99b
          Stem-loop Accession # MI0031246
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036582
          miRBase ID: tch-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306774_pri
          TaqMan™ Pri-miRNA Assay : Rn03465992_pri
          TaqMan™ Pri-miRNA Assay : Hs03295173_pri
          Ambion® Pre-miR™ miRNA Precursor : PM11021
          TaqMan™ MicroRNA Assay : 000436
          mirVana® miRNA inhibitor : MH11021
          mirVana® miRNA mimic : MC11021
          TaqMan™ Advanced miRNA Assay : rno481284_mir
          TaqMan™ Advanced miRNA Assay : 478343_mir
          TaqMan™ Advanced miRNA Assay : mmu481284_mir


          miRBase Alias:

          Human: hsa-miR-99b (v17)
          Mouse: mmu-miR-99b (v17)
          Rat: rno-miR-99b (v18)
          Macaca mulatta: mml-miR-99b (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline