Search
Search

| Assay Name: | hsa-miR-29c-5p |
|---|---|
| miRBase Accession Number: | MI0000735 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGACCGAUUUCUCCUGGUGUUC |
| Species: | Human, Mouse, Rat, Pteropus alecto |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UGACCGAUUUCUCCUGGUGUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-29c-5p | MIMAT0004673 |
| Mouse | mmu-miR-29c-5p | MIMAT0004632 |
| Rat | rno-miR-29c-5p | MIMAT0003154 |
| Pteropus alecto | pal-miR-29c-5p | MIMAT0040197 |