Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12371
          Assay Name: hsa-miR-16-1-3p
          miRBase Accession Number: MI0000070
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 50048973 - 50049061 [-] on Build GRCh38
          Mature miRNA Sequence: CCAGUAUUAACUGUGCUGCUGA
          Species: Human, Alligator mississippiensis, Chrysemys picta, Taeniopygia guttata
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12371
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          CCAGUAUUAACUGUGCUGCUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-16-1-3p MIMAT0004489
          Alligator mississippiensis ami-miR-16a-3p MIMAT0038043
          Chrysemys picta cpi-miR-16a-3p MIMAT0037635
          Taeniopygia guttata tgu-miR-16a-3p MIMAT0014647

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-16-1
          Stem-loop Accession # MI0000070
          Stem-loop Sequence
          GUCAGCAGUGCCUUAGCAGCACGUAAAUAUUGGCGUUAAGAUUCUAAAAUUAUCUCCAGUAUUAACUGUGCUGCUGAAGUAAGGUUGAC
          Chromosome Location Chr. 13 - 50048973 - 50049061 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004489
          miRBase ID: hsa-miR-16-1-3p
          Mature miRNA Sequence: CCAGUAUUAACUGUGCUGCUGA
          Chromosome Location: Chr. 13 - 50048973 - 50049061 [-] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-16a-1
          Stem-loop Accession # MI0029669
          Stem-loop Sequence
          AUGUCAAUCGUACCUUAGCAGCACGUAAAUAUUGGUGUUAAGAUUCUGUAAAUAUCUCCAGUAUUAACUGUGCUGCUGAAGUAAGUUUGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038043
          miRBase ID: ami-miR-16a-3p
          Mature miRNA Sequence: CCAGUAUUAACUGUGCUGCUGA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-16a
          Stem-loop Accession # MI0029400
          Stem-loop Sequence
          CAGCAGUACAUUAGCAGCACGUAAAUAUUGGCGUUAACUCUGUAAAUAUCUCCAGUAUUAACUGUGCUGCUGAAGUAAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037635
          miRBase ID: cpi-miR-16a-3p
          Mature miRNA Sequence: CCAGUAUUAACUGUGCUGCUGA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-16a
          Stem-loop Accession # MI0013811
          Stem-loop Sequence
          UGUCAUACUUUAGCAGCACGUAAAUAUUGGUGUUAAGACUGUAAAUAUCUCCAGUAUUAACUGUGCUGCUGAAGUAAGGCUAGCCACUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014647
          miRBase ID: tgu-miR-16a-3p
          Mature miRNA Sequence: CCAGUAUUAACUGUGCUGCUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302582_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12371
          TaqMan™ MicroRNA Assay : 002420
          mirVana® miRNA inhibitor : MH12371
          mirVana® miRNA mimic : MC12371
          TaqMan™ Advanced miRNA Assay : 478727_mir


          miRBase Alias:

          Human: hsa-miR-16-1* (v17)
          Trabulsiella guamensis ATCC 49490: tgu-miR-16a* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline