Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12588
          Assay Name: rno-miR-135a-3p
          miRBase Accession Number: MI0000908
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 32894877 - 32894976 [-] on Build Rnor_6.0
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Species: Rat, Chrysemys picta, Dog, Eptesicus fuscus, Pteropus alecto, Python bivittatus
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12588
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000028, mir-135

          Mature miRNA Sequence:

          UGUAGGGAUGGAAGCCAUGAAA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-135a-3p MIMAT0004732
          Chrysemys picta cpi-miR-135-2-3p MIMAT0037756
          Dog cfa-miR-135a-3p MIMAT0006628
          Eptesicus fuscus efu-miR-135 MIMAT0034970
          Pteropus alecto pal-miR-135-3p MIMAT0039986
          Python bivittatus pbv-miR-135-2-3p MIMAT0038943

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-135a
          Stem-loop Accession # MI0000908
          Stem-loop Sequence
          AGAUAAAUUCACUCUAGUGCUUUAUGGCUUUUUAUUCCUAUGUGAUCGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAUUCA
          Chromosome Location Chr. 7 - 32894877 - 32894976 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004732
          miRBase ID: rno-miR-135a-3p
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location: Chr. 7 - 32894877 - 32894976 [-] on Build Rnor_6.0

          Chrysemys picta

          Stem-loop ID cpi-mir-135-2
          Stem-loop Accession # MI0029477
          Stem-loop Sequence
          AAUUCACUCUAGUGUUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037756
          miRBase ID: cpi-miR-135-2-3p
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-135a-2
          Stem-loop Accession # MI0008025
          Stem-loop Sequence
          UUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006628
          miRBase ID: cfa-miR-135a-3p
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-135
          Stem-loop Accession # MI0028655
          Stem-loop Sequence
          CUCUAGUGCUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0034970
          miRBase ID: efu-miR-135
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-135-1
          Stem-loop Accession # MI0032434
          Stem-loop Sequence
          UAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039986
          miRBase ID: pal-miR-135-3p
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-135-2
          Stem-loop Accession # MI0030233
          Stem-loop Sequence
          UUCACUCUAGUGUUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038943
          miRBase ID: pbv-miR-135-2-3p
          Mature miRNA Sequence: UGUAGGGAUGGAAGCCAUGAAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465305_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12588
          TaqMan™ MicroRNA Assay : 002075
          mirVana® miRNA inhibitor : MH12588
          mirVana® miRNA mimic : MC12588
          TaqMan™ Advanced miRNA Assay : rno481322_mir


          miRBase Alias:

          Rat: rno-miR-135a* (v18)
          Citrobacter farmeri: cfa-miR-135 (v12)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline