Search
Search

| Assay Name: | hsa-miR-345-5p |
|---|---|
| miRBase Accession Number: | MI0000825 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUGACUCCUAGUCCAGGGCUC |
| Species: | Human, Nomascus leucogenys, Pan troglodytes, Pongo pygmaeus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000189, mir-345 |
Mature miRNA Sequence: | GCUGACUCCUAGUCCAGGGCUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-345-5p | MIMAT0000772 |
| Nomascus leucogenys | nle-miR-345 | MIMAT0049378 |
| Pan troglodytes | ptr-miR-345 | MIMAT0008111 |
| Pongo pygmaeus | ppy-miR-345 | MIMAT0015846 |