Search
Search

| Assay Name: | rno-miR-98-3p |
|---|---|
| miRBase Accession Number: | MI0000882 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUACAACUUACUACUUUCC |
| Species: | Rat, Alligator mississippiensis, Python bivittatus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAACUUACUACUUUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-98-3p | MIMAT0017111 |
| Alligator mississippiensis | ami-miR-98-3p | MIMAT0038109 |
| Python bivittatus | pbv-miR-98-3p | MIMAT0038894 |