Search
Search

| Assay Name: | mmu-miR-184-5p |
|---|---|
| miRBase Accession Number: | MI0000226 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUUAUCACUUUUCCAGCCAGC |
| Species: | Mouse, Anolis carolinensis, Pteropus alecto |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000059, mir-184 |
Mature miRNA Sequence: | CCUUAUCACUUUUCCAGCCAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-184-5p | MIMAT0022690 |
| Anolis carolinensis | aca-miR-184-5p | MIMAT0021813 |
| Pteropus alecto | pal-miR-184-5p | MIMAT0040164 |