Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10206
          Assay Name: hsa-miR-21-5p
          miRBase Accession Number: MI0000077
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 59841266 - 59841337 [+] on Build GRCh38
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Species: Human, Mouse, Rat, Anolis carolinensis, Ateles geoffroyi, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dog, Gorilla gorilla, Horse, Macaca nemestrina, Monodelphis domestica, Ophiophagus hannah, Ornithorhynchus anatinus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10206
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000060, mir-21

          Mature miRNA Sequence:

          UAGCUUAUCAGACUGAUGUUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-21-5p MIMAT0000076
          Mouse mmu-miR-21a-5p MIMAT0000530
          Rat rno-miR-21-5p MIMAT0000790
          Anolis carolinensis aca-miR-21-5p MIMAT0021865
          Ateles geoffroyi age-miR-21 MIMAT0002325
          Callithrix jacchus cja-miR-21 MIMAT0039441
          View More Rows
          Chicken gga-miR-21-5p MIMAT0003774
          Chrysemys picta cpi-miR-21-5p MIMAT0037651
          Columba livia cli-miR-21-5p MIMAT0038436
          Cricetulus griseus cgr-miR-21-5p MIMAT0004417
          Dog cfa-miR-21 MIMAT0006741
          Gorilla gorilla ggo-miR-21 MIMAT0002322
          Horse eca-miR-21 MIMAT0013029
          Macaca nemestrina mne-miR-21 MIMAT0002324
          Monodelphis domestica mdo-miR-21-5p MIMAT0004091
          Ophiophagus hannah oha-miR-21-5p MIMAT0036824
          Ornithorhynchus anatinus oan-miR-21-5p MIMAT0007160
          Pan paniscus ppa-miR-21 MIMAT0002326
          Pan troglodytes ptr-miR-21 MIMAT0002321
          Pig ssc-miR-21-5p MIMAT0002165
          Pongo pygmaeus ppy-miR-21 MIMAT0002323
          Pteropus alecto pal-miR-21-5p MIMAT0039795
          Rhesus monkey mml-miR-21-5p MIMAT0002320
          Taeniopygia guttata tgu-miR-21-5p MIMAT0014527
          Tupaia chinensis tch-miR-21-5p MIMAT0036592

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-21
          Stem-loop Accession # MI0000077
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location Chr. 17 - 59841266 - 59841337 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000076
          miRBase ID: hsa-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: Chr. 17 - 59841266 - 59841337 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-21a
          Stem-loop Accession # MI0000569
          Stem-loop Sequence
          UGUACCACCUUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location Chr. 11 - 86584067 - 86584158 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000530
          miRBase ID: mmu-miR-21a-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: Chr. 11 - 86584067 - 86584158 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-21
          Stem-loop Accession # MI0000850
          Stem-loop Sequence
          UGUACCACCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location Chr. 10 - 73902210 - 73902301 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000790
          miRBase ID: rno-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: Chr. 10 - 73902210 - 73902301 [-] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-21
          Stem-loop Accession # MI0018814
          Stem-loop Sequence
          CGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUGUCAUGGCAACAACAGUCGGUAGGCUGUCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021865
          miRBase ID: aca-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-21
          Stem-loop Accession # MI0002626
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002325
          miRBase ID: age-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-21
          Stem-loop Accession # MI0031964
          Stem-loop Sequence
          GUACCACCUUGUCAGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039441
          miRBase ID: cja-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-21
          Stem-loop Accession # MI0004994
          Stem-loop Sequence
          UGUACCAUCCUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUCUCAUGGCAACAACAGUCGGUAGGCUGUCUGACAUUUUGGUAUCUCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003774
          miRBase ID: gga-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-21
          Stem-loop Accession # MI0029409
          Stem-loop Sequence
          CUGUACCAUCCUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUCUCAUGGCAACAACAGUCGGUAGGCUGUCUGACAUUUUGGUAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037651
          miRBase ID: cpi-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-21
          Stem-loop Accession # MI0029920
          Stem-loop Sequence
          UCCUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUCUCAUGGCAACAACAGUCGGUAGGCUGUCUGACAUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038436
          miRBase ID: cli-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-21
          Stem-loop Accession # MI0005725
          Stem-loop Sequence
          UGUACCACCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004417
          miRBase ID: cgr-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-21
          Stem-loop Accession # MI0008165
          Stem-loop Sequence
          UAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006741
          miRBase ID: cfa-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-21
          Stem-loop Accession # MI0002623
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002322
          miRBase ID: ggo-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-21
          Stem-loop Accession # MI0012776
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013029
          miRBase ID: eca-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-21
          Stem-loop Accession # MI0002625
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002324
          miRBase ID: mne-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-21
          Stem-loop Accession # MI0005275
          Stem-loop Sequence
          UGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUCUCAUGGCAACAGCAGUCGAUGAGCUGUCUGACAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004091
          miRBase ID: mdo-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-21
          Stem-loop Accession # MI0031411
          Stem-loop Sequence
          CUUUCUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUGUCAUGGCAACAACAGUCGGUAGGCUGUCUGACAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036824
          miRBase ID: oha-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-21
          Stem-loop Accession # MI0006883
          Stem-loop Sequence
          AUCUCCACGACUGGACCAUCCUAUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUAGAUCUCCUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUCUGUCAUCCGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007160
          miRBase ID: oan-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-21
          Stem-loop Accession # MI0002627
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002326
          miRBase ID: ppa-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-21
          Stem-loop Accession # MI0002622
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002321
          miRBase ID: ptr-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-21
          Stem-loop Accession # MI0002459
          Stem-loop Sequence
          UGUACCACCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002165
          miRBase ID: ssc-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-21
          Stem-loop Accession # MI0002624
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002323
          miRBase ID: ppy-miR-21
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-21
          Stem-loop Accession # MI0032333
          Stem-loop Sequence
          UAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039795
          miRBase ID: pal-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-21
          Stem-loop Accession # MI0002621
          Stem-loop Sequence
          UGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACACCAGUCGAUGGGCUGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002320
          miRBase ID: mml-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-21
          Stem-loop Accession # MI0013739
          Stem-loop Sequence
          UCCUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGGAUCUCAUGGCAACAACAGUCGGUAGGCUGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014527
          miRBase ID: tgu-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-21
          Stem-loop Accession # MI0031258
          Stem-loop Sequence
          UAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036592
          miRBase ID: tch-miR-21-5p
          Mature miRNA Sequence: UAGCUUAUCAGACUGAUGUUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302625_pri
          TaqMan™ Pri-miRNA Assay : Rn03464993_pri
          TaqMan™ Pri-miRNA Assay : Mm03306822_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10206
          TaqMan™ MicroRNA Assay : 000397
          mirVana® miRNA inhibitor : MH10206
          mirVana® miRNA mimic : MC10206
          TaqMan™ Advanced miRNA Assay : 477975_mir
          TaqMan™ Advanced miRNA Assay : mmu482709_mir
          TaqMan™ Advanced miRNA Assay : rno481342_mir


          miRBase Alias:

          Human: hsa-miR-21 (v17)
          Mouse: mmu-miR-21-5p (v18)
          Mouse: mmu-miR-21 (v17)
          Rat: rno-miR-21 (v18)
          Aeromonas caviae Ae398: aca-miR-21 (v18)
          Comamonas granuli NBRC 101663: cgr-miR-21 (v18)
          Gallus gallus: gga-miR-21 (v19)
          Macaca mulatta: mml-miR-21 (v19)
          Monodelphis domestica: mdo-miR-21 (v19)
          Ochrobactrum anthropi: oan-miR-21 (v18)
          Staphylococcus schleiferi: ssc-miR-21 (v21)
          Trabulsiella guamensis ATCC 49490: tgu-miR-21 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline