Search
Search

| Assay Name: | hsa-miR-30b-5p |
|---|---|
| miRBase Accession Number: | MI0000441 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUAAACAUCCUACACUCAGCU |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Goat, Guinea pig, Horse, Ictalurus punctatus, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Ophiophagus hannah, Oreochromis niloticus, Ornithorhynchus anatinus, Otolemur garnettii, Pig, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Tupaia chinensis, Xenopus laevis, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCUACACUCAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30b-5p | MIMAT0000420 |
| Mouse | mmu-miR-30b-5p | MIMAT0000130 |
| Rat | rno-miR-30b-5p | MIMAT0000806 |
| Alligator mississippiensis | ami-miR-30b-5p | MIMAT0038079 |
| Anolis carolinensis | aca-miR-30b-5p | MIMAT0021922 |
| Astatotilapia burtoni | abu-miR-30d | MIMAT0042084 |
| View More Rows | ||
| Bovine | bta-miR-30b-5p | MIMAT0003547 |
| Callithrix jacchus | cja-miR-30b | MIMAT0039502 |
| Chicken | gga-miR-30b-5p | MIMAT0001130 |
| Chrysemys picta | cpi-miR-30b-5p | MIMAT0037676 |
| Cricetulus griseus | cgr-miR-30b-5p | MIMAT0023896 |
| Cyprinus carpio | ccr-miR-30b | MIMAT0026289 |
| Dasypus novemcinctus | dno-miR-30b-5p | MIMAT0047558 |
| Dog | cfa-miR-30b | MIMAT0006617 |
| Fugu rubripes | fru-miR-30b | MIMAT0002947 |
| Gadus morhua | gmo-miR-30c-5p | MIMAT0044142 |
| Goat | chi-miR-30b-5p | MIMAT0036123 |
| Guinea pig | cpo-miR-30b-5p | MIMAT0046905 |
| Horse | eca-miR-30b | MIMAT0013005 |
| Ictalurus punctatus | ipu-miR-30b | MIMAT0029497 |
| Metriaclima zebra | mze-miR-30d | MIMAT0042339 |
| Monodelphis domestica | mdo-miR-30b-5p | MIMAT0028550 |
| Neolamprologus brichardi | nbr-miR-30d | MIMAT0042527 |
| Nomascus leucogenys | nle-miR-30b | MIMAT0049368 |
| Ophiophagus hannah | oha-miR-30b-5p | MIMAT0036890 |
| Oreochromis niloticus | oni-miR-30d | MIMAT0042707 |
| Ornithorhynchus anatinus | oan-miR-30b-5p | MIMAT0007043 |
| Otolemur garnettii | oga-miR-30b | MIMAT0049616 |
| Pig | ssc-miR-30b-5p | MIMAT0007756 |
| Pongo pygmaeus | ppy-miR-30b | MIMAT0015736 |
| Pteropus alecto | pal-miR-30b-5p | MIMAT0039945 |
| Pundamilia nyererei | pny-miR-30d | MIMAT0042905 |
| Python bivittatus | pbv-miR-30b-5p | MIMAT0038872 |
| Rabbit | ocu-miR-30b-5p | MIMAT0048152 |
| Salmo salar | ssa-miR-30e-5p | MIMAT0032611 |
| Taeniopygia guttata | tgu-miR-30e | MIMAT0025411 |
| Tetraodon nigroviridis | tni-miR-30b | MIMAT0002948 |
| Tupaia chinensis | tch-miR-30b-5p | MIMAT0036503 |
| Xenopus laevis | xla-miR-30b-5p | MIMAT0046570 |
| Xenopus tropicalis | xtr-miR-30b | MIMAT0003576 |
| Zebrafish | dre-miR-30b | MIMAT0001804 |