Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11142
          Assay Name: hsa-miR-455-3p
          miRBase Accession Number: MI0003513
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 114209434 - 114209529 [+] on Build GRCh38
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Species: Human, Anolis carolinensis, Gorilla gorilla, Ophiophagus hannah, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11142
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000129, mir-455

          Mature miRNA Sequence:

          GCAGUCCAUGGGCAUAUACAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-455-3p MIMAT0004784
          Anolis carolinensis aca-miR-455-3p MIMAT0021965
          Gorilla gorilla ggo-miR-455 MIMAT0024222
          Ophiophagus hannah oha-miR-455-3p MIMAT0036932
          Pan troglodytes ptr-miR-455 MIMAT0008157
          Pig ssc-miR-455-3p MIMAT0013960
          View More Rows
          Pongo pygmaeus ppy-miR-455-3p MIMAT0015900
          Python bivittatus pbv-miR-455-3p MIMAT0039058
          Rhesus monkey mml-miR-455-3p MIMAT0006340

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-455
          Stem-loop Accession # MI0003513
          Stem-loop Sequence
          UCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location Chr. 9 - 114209434 - 114209529 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004784
          miRBase ID: hsa-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: Chr. 9 - 114209434 - 114209529 [+] on Build GRCh38

          Anolis carolinensis

          Stem-loop ID aca-mir-455
          Stem-loop Accession # MI0018871
          Stem-loop Sequence
          UCCCUGGCGUGGGGGUAUGUGCCCUUGGACUACAUCGUGGGAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGUUUAUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021965
          miRBase ID: aca-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-455
          Stem-loop Accession # MI0020769
          Stem-loop Sequence
          GCGAGCGCCUGCGCCCUUCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024222
          miRBase ID: ggo-miR-455
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-455
          Stem-loop Accession # MI0031473
          Stem-loop Sequence
          ACUGGUCCCUGGUGUGAGGGUAUGUGCCCUUGGACUACAUCGUGGAAAAUAUCACCAUGCAGUCCAUGGGCAUAUACACUUGCCCCAAGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036932
          miRBase ID: oha-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-455
          Stem-loop Accession # MI0008679
          Stem-loop Sequence
          CCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008157
          miRBase ID: ptr-miR-455
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-455
          Stem-loop Accession # MI0013170
          Stem-loop Sequence
          UGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013960
          miRBase ID: ssc-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-455
          Stem-loop Accession # MI0014955
          Stem-loop Sequence
          UCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015900
          miRBase ID: ppy-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-455
          Stem-loop Accession # MI0030315
          Stem-loop Sequence
          GAUCCCUGGUGUGAGGGUAUGUGCCCUUGGACUACAUCGUGGAAAAUAUCACCAUGCAGUCCAUGGGCAUAUACACUUGCCCCAAGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039058
          miRBase ID: pbv-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-455
          Stem-loop Accession # MI0007756
          Stem-loop Sequence
          UCCCUGGCGUGAGGGUAUGUGCCUUUGGACUACAUCGUGGAAGCCAGCACCAUGCAGUCCAUGGGCAUAUACACUUGCCUCAAGGCCUAUGUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006340
          miRBase ID: mml-miR-455-3p
          Mature miRNA Sequence: GCAGUCCAUGGGCAUAUACAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03294041_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11142
          TaqMan™ MicroRNA Assay : 002244
          mirVana® miRNA inhibitor : MH11142
          mirVana® miRNA mimic : MC11142
          TaqMan™ Advanced miRNA Assay : 478112_mir


          miRBase Alias:

          Staphylococcus schleiferi: ssc-miR-455 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline