Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11714
          Assay Name: hsa-miR-200c-3p
          miRBase Accession Number: MI0000650
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 6963699 - 6963766 [+] on Build GRCh38
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Species: Human, Mouse, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Eptesicus fuscus, Goat, Guinea pig, Horse, Pan troglodytes, Pteropus alecto, Rabbit, Tupaia chinensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11714
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          UAAUACUGCCGGGUAAUGAUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-200c-3p MIMAT0000617
          Mouse mmu-miR-200c-3p MIMAT0000657
          Bovine bta-miR-200c MIMAT0003823
          Callithrix jacchus cja-miR-200c MIMAT0039426
          Cricetulus griseus cgr-miR-200c MIMAT0023843
          Dog cfa-miR-200c MIMAT0006664
          View More Rows
          Eptesicus fuscus efu-miR-200c MIMAT0035096
          Goat chi-miR-200c MIMAT0036047
          Guinea pig cpo-miR-200c-3p MIMAT0047094
          Horse eca-miR-200c MIMAT0012974
          Pan troglodytes ptr-miR-200c MIMAT0024051
          Pteropus alecto pal-miR-200c-3p MIMAT0039912
          Rabbit ocu-miR-200c-3p MIMAT0048322
          Tupaia chinensis tch-miR-200c-3p MIMAT0036473

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-200c
          Stem-loop Accession # MI0000650
          Stem-loop Sequence
          CCCUCGUCUUACCCAGCAGUGUUUGGGUGCGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGG
          Chromosome Location Chr. 12 - 6963699 - 6963766 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000617
          miRBase ID: hsa-miR-200c-3p
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: Chr. 12 - 6963699 - 6963766 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-200c
          Stem-loop Accession # MI0000694
          Stem-loop Sequence
          CCCUCGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGG
          Chromosome Location Chr. 6 - 124718322 - 124718390 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000657
          miRBase ID: mmu-miR-200c-3p
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: Chr. 6 - 124718322 - 124718390 [-] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-200c
          Stem-loop Accession # MI0005038
          Stem-loop Sequence
          CGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003823
          miRBase ID: bta-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-200c
          Stem-loop Accession # MI0031946
          Stem-loop Sequence
          GGGUGGGGGCCCUCGUCUUACCCAGCAGUGUUUGGGUGCGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGGCCCCUGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039426
          miRBase ID: cja-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-200c
          Stem-loop Accession # MI0020453
          Stem-loop Sequence
          GGUUGGGUAGGGGCCCUCGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGGCCCCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023843
          miRBase ID: cgr-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-200c
          Stem-loop Accession # MI0008070
          Stem-loop Sequence
          CGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006664
          miRBase ID: cfa-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-200c
          Stem-loop Accession # MI0028789
          Stem-loop Sequence
          UGGGGGCCCCCGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGGCCCCCGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035096
          miRBase ID: efu-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-200c
          Stem-loop Accession # MI0030681
          Stem-loop Sequence
          GGGUGGGGGCCCUCGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGGCCCCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036047
          miRBase ID: chi-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-200c
          Stem-loop Accession # MI0038694
          Stem-loop Sequence
          CGUCUUACCCUGCAGUGUUUGGGUGCUGGUUUGGUCUCUAAUACUGCCGGGUAAUGAUGGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047094
          miRBase ID: cpo-miR-200c-3p
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-200c
          Stem-loop Accession # MI0012727
          Stem-loop Sequence
          CCGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012974
          miRBase ID: eca-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-200c
          Stem-loop Accession # MI0020598
          Stem-loop Sequence
          ACCCAAGGUGGGCGGGCUGGGCGGGGGCCCUCGUCUUACCCAGCAGUGUUUGGGUGCGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGAGGCCCCUGUCCCUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024051
          miRBase ID: ptr-miR-200c
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-200c
          Stem-loop Accession # MI0032396
          Stem-loop Sequence
          CGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039912
          miRBase ID: pal-miR-200c-3p
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-200c
          Stem-loop Accession # MI0039340
          Stem-loop Sequence
          CGUCUUACCCAGCAGUGUUUGGGUGCUGGUUGGGAGUCUCUAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048322
          miRBase ID: ocu-miR-200c-3p
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-200c
          Stem-loop Accession # MI0031132
          Stem-loop Sequence
          CGUCUUACCCAGCAGUGUUUGGGUGUUGCUUGGCAUUCUCUAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036473
          miRBase ID: tch-miR-200c-3p
          Mature miRNA Sequence: UAAUACUGCCGGGUAAUGAUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307102_pri
          TaqMan™ Pri-miRNA Assay : Hs03303157_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11714
          TaqMan™ MicroRNA Assay : 002300
          mirVana® miRNA inhibitor : MH11714
          mirVana® miRNA mimic : MC11714
          TaqMan™ Advanced miRNA Assay : 478351_mir
          TaqMan™ Advanced miRNA Assay : mmu482938_mir


          miRBase Alias:

          Human: hsa-miR-200c (v17)
          Mouse: mmu-miR-200c (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline