Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11717
          Assay Name: hsa-miR-191-5p
          miRBase Accession Number: MI0000465
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 49020618 - 49020709 [-] on Build GRCh38
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Species: Human, Mouse, Rat, Anolis carolinensis, Bovine, Callithrix jacchus, Chicken, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ophiophagus hannah, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Saimiri boliviensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11717
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000194, mir-191

          Mature miRNA Sequence:

          CAACGGAAUCCCAAAAGCAGCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-191-5p MIMAT0000440
          Mouse mmu-miR-191-5p MIMAT0000221
          Rat rno-miR-191a-5p MIMAT0000866
          Anolis carolinensis aca-miR-191-5p MIMAT0021824
          Bovine bta-miR-191 MIMAT0003819
          Callithrix jacchus cja-miR-191 MIMAT0039392
          View More Rows
          Chicken gga-miR-191-5p MIMAT0049917
          Cricetulus griseus cgr-miR-191-5p MIMAT0023822
          Dasypus novemcinctus dno-miR-191-5p MIMAT0047717
          Daubentonia madagascariensis dma-miR-191 MIMAT0049295
          Guinea pig cpo-miR-191-5p MIMAT0047066
          Horse eca-miR-191a MIMAT0013079
          Microcebus murinus mmr-miR-191 MIMAT0049213
          Nomascus leucogenys nle-miR-191 MIMAT0049457
          Ophiophagus hannah oha-miR-191-5p MIMAT0036777
          Otolemur garnettii oga-miR-191 MIMAT0049685
          Pan paniscus ppa-miR-191 MIMAT0049078
          Pan troglodytes ptr-miR-191 MIMAT0008056
          Papio hamadryas pha-miR-191 MIMAT0049566
          Pig ssc-miR-191 MIMAT0013876
          Pongo pygmaeus ppy-miR-191 MIMAT0015785
          Python bivittatus pbv-miR-191-5p MIMAT0038987
          Rabbit ocu-miR-191-5p MIMAT0036335
          Rhesus monkey mml-miR-191-5p MIMAT0006223
          Saimiri boliviensis sbo-miR-191 MIMAT0049484

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-191
          Stem-loop Accession # MI0000465
          Stem-loop Sequence
          CGGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCCUGCUCUCCUGCCU
          Chromosome Location Chr. 3 - 49020618 - 49020709 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000440
          miRBase ID: hsa-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: Chr. 3 - 49020618 - 49020709 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-191
          Stem-loop Accession # MI0000233
          Stem-loop Sequence
          AGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCACUUGGAUUUCGUUCCCUGCU
          Chromosome Location Chr. 9 - 108568319 - 108568392 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000221
          miRBase ID: mmu-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: Chr. 9 - 108568319 - 108568392 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-191a
          Stem-loop Accession # MI0000934
          Stem-loop Sequence
          GGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCACUUGGAUUUCGUUCCCUGCUCUCCUGCCU
          Chromosome Location Chr. 8 - 117354364 - 117354454 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000866
          miRBase ID: rno-miR-191a-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: Chr. 8 - 117354364 - 117354454 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-191
          Stem-loop Accession # MI0018787
          Stem-loop Sequence
          GUUUGAUAGUGGGCAACGGAAUCCCAAAAGCAGCUGUUUUCCUCUUAAAGUUCCAGCUGCACUUGGAUUUCGUUCCUUGCUUUCCUACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021824
          miRBase ID: aca-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-191
          Stem-loop Accession # MI0005034
          Stem-loop Sequence
          GGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCCUGCUCUCCUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003819
          miRBase ID: bta-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-191
          Stem-loop Accession # MI0031911
          Stem-loop Sequence
          GCCAACGGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCCUGCUCUUCCGCCUGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039392
          miRBase ID: cja-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-191
          Stem-loop Accession # MI0040675
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUCCCCGCAGCGCCCAGCUGCCCUGGGAUUUCGUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049917
          miRBase ID: gga-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-191
          Stem-loop Accession # MI0020437
          Stem-loop Sequence
          GACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCGAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCCUGCUCUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023822
          miRBase ID: cgr-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-191
          Stem-loop Accession # MI0039022
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047717
          miRBase ID: dno-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-191
          Stem-loop Accession # MI0040030
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049295
          miRBase ID: dma-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-191
          Stem-loop Accession # MI0038679
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUUGAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047066
          miRBase ID: cpo-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-191a
          Stem-loop Accession # MI0012825
          Stem-loop Sequence
          AGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCCUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013079
          miRBase ID: eca-miR-191a
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-191
          Stem-loop Accession # MI0039938
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049213
          miRBase ID: mmr-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-191
          Stem-loop Accession # MI0040209
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049457
          miRBase ID: nle-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-191
          Stem-loop Accession # MI0031386
          Stem-loop Sequence
          GAAUGGGCAACGGAAUCCCAAAAGCAGCUGUUUUGCUCUGUUUCCCAGCUGCACUUGGAUUCCGUUCCUUGCUCUUUUGCCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036777
          miRBase ID: oha-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-191
          Stem-loop Accession # MI0040469
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049685
          miRBase ID: oga-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-191
          Stem-loop Accession # MI0039786
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049078
          miRBase ID: ppa-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-191
          Stem-loop Accession # MI0008564
          Stem-loop Sequence
          CGGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCCUGCUCUCCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008056
          miRBase ID: ptr-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-191
          Stem-loop Accession # MI0040336
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049566
          miRBase ID: pha-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-191
          Stem-loop Accession # MI0013095
          Stem-loop Sequence
          ACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCACUUGGAUUUCGUUCCCUGCUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013876
          miRBase ID: ssc-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-191
          Stem-loop Accession # MI0014853
          Stem-loop Sequence
          CGGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCCUGCUCUCCUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015785
          miRBase ID: ppy-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-191
          Stem-loop Accession # MI0030264
          Stem-loop Sequence
          UGGGCAACGGAAUCCCAAAAGCAGCUGUUUUCCUCUGAGUCUUCCAGCUGCACUUGGAUUCCGUUCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038987
          miRBase ID: pbv-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-191
          Stem-loop Accession # MI0030998
          Stem-loop Sequence
          CGGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUUCCCUGCUUUCCUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036335
          miRBase ID: ocu-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-191
          Stem-loop Accession # MI0007653
          Stem-loop Sequence
          CGGCUGGACAGCGGGCAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCCUGCUCUCCUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006223
          miRBase ID: mml-miR-191-5p
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-191
          Stem-loop Accession # MI0040244
          Stem-loop Sequence
          CAACGGAAUCCCAAAAGCAGCUGUUGUCUCCAGAGCAUUCCAGCUGCGCUUGGAUUUCGUCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049484
          miRBase ID: sbo-miR-191
          Mature miRNA Sequence: CAACGGAAUCCCAAAAGCAGCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303702_pri
          TaqMan™ Pri-miRNA Assay : Rn03465415_pri
          TaqMan™ Pri-miRNA Assay : Mm03306480_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11717
          TaqMan™ MicroRNA Assay : 002299
          mirVana® miRNA inhibitor : MH11717
          mirVana® miRNA mimic : MC11717
          TaqMan™ Advanced miRNA Assay : rno480971_mir
          TaqMan™ Advanced miRNA Assay : mmu481584_mir
          TaqMan™ Advanced miRNA Assay : 477952_mir


          miRBase Alias:

          Human: hsa-miR-191 (v17)
          Mouse: mmu-miR-191 (v17)
          Rat: rno-miR-191 (v18)
          Aeromonas caviae Ae398: aca-miR-191 (v18)
          Enterovirus CA55-1988: eca-miR-191 (v20)
          Macaca mulatta: mml-miR-191 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline