Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM14801
          Assay Name: mmu-miR-96-3p
          miRBase Accession Number: MI0000583
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 30169446 - 30169551 [-] on Build GRCm38
          Mature miRNA Sequence: CAAUCAUGUGUAGUGCCAAUAU
          Species: Mouse, Ornithorhynchus anatinus, Salmo salar
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM14801
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000072, mir-96

          Mature miRNA Sequence:

          CAAUCAUGUGUAGUGCCAAUAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-96-3p MIMAT0017021
          Ornithorhynchus anatinus oan-miR-96-3p MIMAT0006804
          Salmo salar ssa-miR-96-3p MIMAT0032717

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-96
          Stem-loop Accession # MI0000583
          Stem-loop Sequence
          CCAGUACCAUCUGCUUGGCCGAUUUUGGCACUAGCACAUUUUUGCUUGUGUCUCUCCGCUGUGAGCAAUCAUGUGUAGUGCCAAUAUGGGAAAAGCGGGCUGCUGC
          Chromosome Location Chr. 6 - 30169446 - 30169551 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017021
          miRBase ID: mmu-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGUAGUGCCAAUAU
          Chromosome Location: Chr. 6 - 30169446 - 30169551 [-] on Build GRCm38

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-96
          Stem-loop Accession # MI0006666
          Stem-loop Sequence
          GUGGAGAUGUCUGCUUGGCCUCUUUUGGCACUAGCACAUUUUUGCUUUUGUCUCUCUGCUCUGAGCAAUCAUGUGUAGUGCCAAUAUGGGAAAAGCUGGAAUUGUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006804
          miRBase ID: oan-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGUAGUGCCAAUAU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-96-1
          Stem-loop Accession # MI0026765
          Stem-loop Sequence
          UUUGGCACUAGCACAUUUUUGCUUCUGUAUAUAUGCUUUGAGCAAUCAUGUGUAGUGCCAAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032717
          miRBase ID: ssa-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGUAGUGCCAAUAU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-96-2
          Stem-loop Accession # MI0026766
          Stem-loop Sequence
          UUUGGCACUAGCACAUUUUUGCUUCUGUAUAUAUCUGUUUGAGCAAUCAUGUGUAGUGCCAAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032717
          miRBase ID: ssa-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGUAGUGCCAAUAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306426_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM14801
          TaqMan™ MicroRNA Assay : 006641_mat
          mirVana® miRNA inhibitor : MH14801
          mirVana® miRNA mimic : MC14801
          TaqMan™ Advanced miRNA Assay : mmu482913_mir


          miRBase Alias:

          Mouse: mmu-miR-96* (v17)
          Ochrobactrum anthropi: oan-miR-96* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline