Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM19034
          Assay Name: hsa-miR-3120-3p
          miRBase Accession Number: MI0014136
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 172138808 - 172138888 [+] on Build GRCh38
          Mature miRNA Sequence: CACAGCAAGUGUAGACAGGCA
          Species: Human, Rat, Artibeus jamaicensis, Bovine, Chicken
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM19034
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000062, mir-214

          Mature miRNA Sequence:

          CACAGCAAGUGUAGACAGGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3120-3p MIMAT0014982
          Rat rno-miR-3120 MIMAT0017900
          Artibeus jamaicensis aja-miR-3120 MIMAT0031146
          Bovine bta-miR-3120 MIMAT0024572
          Chicken gga-miR-214b-3p MIMAT0050063

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3120
          Stem-loop Accession # MI0014136
          Stem-loop Sequence
          GUCAUGUGACUGCCUGUCUGUGCCUGCUGUACAGGUGAGCGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGACACAUGAC
          Chromosome Location Chr. 1 - 172138808 - 172138888 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0014982
          miRBase ID: hsa-miR-3120-3p
          Mature miRNA Sequence: CACAGCAAGUGUAGACAGGCA
          Chromosome Location: Chr. 1 - 172138808 - 172138888 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-3120
          Stem-loop Accession # MI0015476
          Stem-loop Sequence
          GGUUGUCAUUCAGGCUGGGUUGUCAUGUGACUGCCUGUCUGUGCCUGCUGUACAGGUGAGCGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGACACAUGACAACUCUGUCCAUCCA
          Chromosome Location Chr. 13 - 80130909 - 80131025 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017900
          miRBase ID: rno-miR-3120
          Mature miRNA Sequence: CACAGCAAGUGUAGACAGGCA
          Chromosome Location: Chr. 13 - 80130909 - 80131025 [-] on Build Rnor_6.0

          Artibeus jamaicensis

          Stem-loop ID aja-mir-3120
          Stem-loop Accession # MI0024368
          Stem-loop Sequence
          GUCAUGUGACUGCCUGUCUGUGCCUGCUGUACAGGUGAGCGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGACACAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031146
          miRBase ID: aja-miR-3120
          Mature miRNA Sequence: CACAGCAAGUGUAGACAGGCA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-3120
          Stem-loop Accession # MI0021116
          Stem-loop Sequence
          GUCAUGUGACUGCCUGUCUGUGCCUGCUGUACAGGUGAGCGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGACACAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024572
          miRBase ID: bta-miR-3120
          Mature miRNA Sequence: CACAGCAAGUGUAGACAGGCA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-214b
          Stem-loop Accession # MI0040752
          Stem-loop Sequence
          CUGCCUGUCUGUGCCUGCUGUACAGGUGAGAGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0050063
          miRBase ID: gga-miR-214b-3p
          Mature miRNA Sequence: CACAGCAAGUGUAGACAGGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302971_pri
          TaqMan™ Pri-miRNA Assay : Rn03465830_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19034
          TaqMan™ MicroRNA Assay : 242355_mat
          mirVana® miRNA inhibitor : MH19034
          mirVana® miRNA mimic : MC19034
          TaqMan™ Advanced miRNA Assay : 478009_mir
          TaqMan™ Advanced miRNA Assay : rno481044_mir


          miRBase Alias:

          Human: hsa-miR-3120 (v16)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline