Search
Search

| Assay Name: | mmu-miR-1264-5p |
|---|---|
| miRBase Accession Number: | MI0004130 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUCCUCAAUAAGUAUUUGUU |
| Species: | Mouse, Guinea pig, Rabbit |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | AGGUCCUCAAUAAGUAUUUGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1264-5p | MIMAT0014802 |
| Guinea pig | cpo-miR-1264-5p | MIMAT0047409 |
| Rabbit | ocu-miR-1264-5p | MIMAT0048589 |