Search
Search

| Assay Name: | hsa-miR-1270 |
|---|---|
| miRBase Accession Number: | MI0006407 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGGAGAUAUGGAAGAGCUGUGU |
| Species: | Human |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000877, mir-1270 |
Mature miRNA Sequence: | CUGGAGAUAUGGAAGAGCUGUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1270 | MIMAT0005924 |