Search
Search

| Assay Name: | hsa-miR-372-3p |
|---|---|
| miRBase Accession Number: | MI0000780 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAGUGCUGCGACAUUUGAGCGU |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000068, mir-290 |
Mature miRNA Sequence: | AAAGUGCUGCGACAUUUGAGCGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-372-3p | MIMAT0000724 |
| Pan troglodytes | ptr-miR-372 | MIMAT0008122 |
| Pongo pygmaeus | ppy-miR-372 | MIMAT0015860 |
| Rhesus monkey | mml-miR-372-3p | MIMAT0006298 |