Search
Search

| Assay Name: | hsa-miR-429 |
|---|---|
| miRBase Accession Number: | MI0001641 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAUACUGUCUGGUAAAACCGU |
| Species: | Human, Callithrix jacchus, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | UAAUACUGUCUGGUAAAACCGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-429 | MIMAT0001536 |
| Callithrix jacchus | cja-miR-429 | MIMAT0039427 |
| Pongo pygmaeus | ppy-miR-429 | MIMAT0015886 |
| Rhesus monkey | mml-miR-429-3p | MIMAT0006325 |