Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10426
          Assay Name: hsa-miR-98-5p
          miRBase Accession Number: MI0000100
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 53556223 - 53556341 [-] on Build GRCh38
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Species: Human, Mouse, Rat, Ateles geoffroyi, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Gorilla gorilla, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Tupaia chinensis, Xenopus laevis, Xenopus tropicalis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10426
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          UGAGGUAGUAAGUUGUAUUGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-98-5p MIMAT0000096
          Mouse mmu-miR-98-5p MIMAT0000545
          Rat rno-miR-98-5p MIMAT0000819
          Ateles geoffroyi age-miR-98 MIMAT0002408
          Bovine bta-miR-98 MIMAT0003809
          Callithrix jacchus cja-miR-98 MIMAT0049302
          View More Rows
          Cricetulus griseus cgr-miR-98 MIMAT0024015
          Dasypus novemcinctus dno-miR-98-5p MIMAT0047592
          Daubentonia madagascariensis dma-miR-98 MIMAT0049292
          Dog cfa-miR-98 MIMAT0006756
          Goat chi-miR-98-5p MIMAT0036310
          Gorilla gorilla ggo-miR-98 MIMAT0002406
          Guinea pig cpo-miR-98-5p MIMAT0046939
          Horse eca-miR-98 MIMAT0013247
          Microcebus murinus mmr-miR-98 MIMAT0049163
          Nomascus leucogenys nle-miR-98 MIMAT0049366
          Ophiophagus hannah oha-miR-98-5p MIMAT0036962
          Ornithorhynchus anatinus oan-miR-98 MIMAT0007178
          Otolemur garnettii oga-miR-98 MIMAT0049654
          Pan paniscus ppa-miR-98 MIMAT0002409
          Pan troglodytes ptr-miR-98 MIMAT0002405
          Papio hamadryas pha-miR-98 MIMAT0049569
          Pig ssc-miR-98 MIMAT0013905
          Pongo pygmaeus ppy-miR-98 MIMAT0002407
          Pteropus alecto pal-miR-98-5p MIMAT0039882
          Python bivittatus pbv-miR-98-5p MIMAT0038893
          Rabbit ocu-miR-98-5p MIMAT0048180
          Rhesus monkey mml-miR-98 MIMAT0002404
          Tupaia chinensis tch-miR-98-5p MIMAT0036570
          Xenopus laevis xla-miR-98-5p MIMAT0046617
          Xenopus tropicalis xtr-miR-98 MIMAT0003581

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-98
          Stem-loop Accession # MI0000100
          Stem-loop Sequence
          AGGAUUCUGCUCAUGCCAGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCCUGGUGUGUGGCAUAUUCA
          Chromosome Location Chr. X - 53556223 - 53556341 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000096
          miRBase ID: hsa-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: Chr. X - 53556223 - 53556341 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-98
          Stem-loop Accession # MI0000586
          Stem-loop Sequence
          CUGCACAUGCUGGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAGUAAGAAGAUAACUAUACAACUUACUACUUUCCUUGGUGUGUGGCAU
          Chromosome Location Chr. X - 151913214 - 151913321 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000545
          miRBase ID: mmu-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: Chr. X - 151913214 - 151913321 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-98
          Stem-loop Accession # MI0000882
          Stem-loop Sequence
          CUGCACAUGCUGGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUAAGAAGAUAACUAUACAACUUACUACUUUCCCUGGUGUGUGGCAU
          Chromosome Location Chr. X - 21583875 - 21583982 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000819
          miRBase ID: rno-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: Chr. X - 21583875 - 21583982 [+] on Build Rnor_6.0

          Ateles geoffroyi

          Stem-loop ID age-mir-98
          Stem-loop Accession # MI0002700
          Stem-loop Sequence
          GUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002408
          miRBase ID: age-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-98
          Stem-loop Accession # MI0005025
          Stem-loop Sequence
          AGGACUCUGCUCAUGCUGGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUUUGAAGAUAACUAUACAACUUACUACUUUCCCUGGUGUGUAGCACAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003809
          miRBase ID: bta-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-98
          Stem-loop Accession # MI0040038
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049302
          miRBase ID: cja-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-98
          Stem-loop Accession # MI0020563
          Stem-loop Sequence
          AUGCUGGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUGGGCCCCAAUAAGAAGAUAACUAUACAACUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024015
          miRBase ID: cgr-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-98
          Stem-loop Accession # MI0038953
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUUAGAAGAUAACUAUGCAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047592
          miRBase ID: dno-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-98
          Stem-loop Accession # MI0040025
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGACCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049292
          miRBase ID: dma-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-98
          Stem-loop Accession # MI0008182
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006756
          miRBase ID: cfa-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-98
          Stem-loop Accession # MI0030844
          Stem-loop Sequence
          AGGACUCUGCUCAUGCUGGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCCUGGUGUGUAGCACAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036310
          miRBase ID: chi-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-98
          Stem-loop Accession # MI0002698
          Stem-loop Sequence
          GUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002406
          miRBase ID: ggo-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-98
          Stem-loop Accession # MI0038609
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAAGGAUUUAAGGCCCCAUUAAGAAGAUAACUAUACGACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046939
          miRBase ID: cpo-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-98
          Stem-loop Accession # MI0012988
          Stem-loop Sequence
          AGGAUUCUGCUCAUGCCAGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCCUGGUGUGUGGCAUAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013247
          miRBase ID: eca-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-98
          Stem-loop Accession # MI0039879
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGAAUAUUAGGCCCCAGUUAGAAGAUAACUAUACAACUUACUACUUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049163
          miRBase ID: mmr-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-98
          Stem-loop Accession # MI0040103
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049366
          miRBase ID: nle-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-98
          Stem-loop Accession # MI0031494
          Stem-loop Sequence
          CUUCUUGCUCCUACCAGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUGUGCCCCAAAUCAGAGAUAACUAUACAACUUACUACUUUCCCUGCGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036962
          miRBase ID: oha-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-98
          Stem-loop Accession # MI0006894
          Stem-loop Sequence
          UCGUAAGCUGUCAGUGUGCCCAUGCUGGGAUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUCUUUUGCCCCAAUUAGCGAUAACUAUACAAUUUACUACUUUCCCCAGUGUGUGGAACGAUCACUGGGUCCUAGUGUAACCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007178
          miRBase ID: oan-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-98
          Stem-loop Accession # MI0040437
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGACCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049654
          miRBase ID: oga-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-98
          Stem-loop Accession # MI0002701
          Stem-loop Sequence
          GUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002409
          miRBase ID: ppa-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-98
          Stem-loop Accession # MI0002697
          Stem-loop Sequence
          AGGAUUCUGCUCAUGCCAGGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCCUGGUGUGUGGCAUAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002405
          miRBase ID: ptr-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-98
          Stem-loop Accession # MI0040340
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049569
          miRBase ID: pha-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-98
          Stem-loop Accession # MI0013122
          Stem-loop Sequence
          GGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013905
          miRBase ID: ssc-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-98
          Stem-loop Accession # MI0002699
          Stem-loop Sequence
          GUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002407
          miRBase ID: ppy-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-98
          Stem-loop Accession # MI0032380
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAAGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039882
          miRBase ID: pal-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-98
          Stem-loop Accession # MI0030200
          Stem-loop Sequence
          GGGUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUGUGCCCCAAAUCAGAGAUAACUAUACAACUUACUACUUUCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038893
          miRBase ID: pbv-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-98
          Stem-loop Accession # MI0039263
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUUUUAGGCCCCAAUAAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048180
          miRBase ID: ocu-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-98
          Stem-loop Accession # MI0002696
          Stem-loop Sequence
          GUGAGGUAGUAAGUUGUAUUGUUGUGGGGUAGGGAUAUUAGGCCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002404
          miRBase ID: mml-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-98
          Stem-loop Accession # MI0031233
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGUUGGGAUUGUAGACCCCAAUUAGAAGAUAACUAUACAACUUACUACUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036570
          miRBase ID: tch-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-98
          Stem-loop Accession # MI0038343
          Stem-loop Sequence
          UGAGGUAGUAAGUUGUAUUGUUGUGGGGGUGUUUCAUUUCCCCAAAUUAGGAGAUAACUAUACAGUUUACUGCCUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046617
          miRBase ID: xla-miR-98-5p
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-98
          Stem-loop Accession # MI0004820
          Stem-loop Sequence
          UCUGCCCUCUUCAGGAUGAGGUAGUAAGUUGUAUUGUUGUGGGGGUUUUUAUUUCCCCAAAUUAGGAGAUAACUAUACAGCUUACUGCCUUCCCUGCAGCAUGGUAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003581
          miRBase ID: xtr-miR-98
          Mature miRNA Sequence: UGAGGUAGUAAGUUGUAUUGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306906_pri
          TaqMan™ Pri-miRNA Assay : Hs03302726_pri
          TaqMan™ Pri-miRNA Assay : Rn03465456_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10426
          TaqMan™ MicroRNA Assay : 000577
          mirVana® miRNA inhibitor : MH10426
          mirVana® miRNA mimic : MC10426
          TaqMan™ Advanced miRNA Assay : rno478590_mir
          TaqMan™ Advanced miRNA Assay : mmu478590_mir
          TaqMan™ Advanced miRNA Assay : 478590_mir


          miRBase Alias:

          Human: hsa-miR-98 (v18)
          Mouse: mmu-miR-98 (v17)
          Rat: rno-miR-98 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline