Search
Search

| Assay Name: | rno-miR-346 |
|---|---|
| miRBase Accession Number: | MI0000633 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCUGCCUGAGUGCCUGCCUCU |
| Species: | Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000188, mir-346 |
Mature miRNA Sequence: | UGUCUGCCUGAGUGCCUGCCUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-346 | MIMAT0000596 |