Search
Search

| Assay Name: | hsa-miR-548c-3p |
|---|---|
| miRBase Accession Number: | MI0003630 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAAAAUCUCAAUUACUUUUGC |
| Species: | Human, Callithrix jacchus, Pan troglodytes |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAAAUCUCAAUUACUUUUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548c-3p | MIMAT0003285 |
| Callithrix jacchus | cja-miR-548j | MIMAT0039602 |
| Pan troglodytes | ptr-miR-548c | MIMAT0008220 |