Search
Search

| Assay Name: | hsa-miR-765 |
|---|---|
| miRBase Accession Number: | MI0005116 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAGGAGAAGGAAGGUGAUG |
| Species: | Human, Pan troglodytes, Pongo pygmaeus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000545, mir-765 |
Mature miRNA Sequence: | UGGAGGAGAAGGAAGGUGAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-765 | MIMAT0003945 |
| Pan troglodytes | ptr-miR-765 | MIMAT0008320 |
| Pongo pygmaeus | ppy-miR-765 | MIMAT0016105 |